Gene Transcripts

Gene Symbol
Kcnb1
Entrez Gene ID
16500
Gene Name
potassium voltage gated channel, Shab-related subfamily, member 1
Gene Aliases
Kcr1-1, Kv2.1, Shab
Chromosome Location
Chr. 2: 167095969 - 167188818 on Build GRCm38
UniGene ID
Mm.205341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008420.4 NP_032446.2 2 1427
GenBank mRNA BC031776.1 - 2 1383
BC051422.1 - 2 1384 6493266
BC061501.1 - 2 1408 4506061
M64228.1 - 2 1232

View Full Product Details