Gene Transcripts
- Gene Symbol
- Vmn1r89
- Entrez Gene ID
- 171260
- Gene Name
- vomeronasal 1 receptor 89
- Gene Aliases
- V1RE8, V1rj2
- Chromosome Location
- Chr. 7: 13219339 - 13220289 on Build GRCm38
- UniGene ID
- Mm.377214
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134226.1 |
NP_598987.1
|
1 |
292 |
|
- Gene Symbol
- Vmn1r86
- Entrez Gene ID
- 100312473
- Gene Name
- vomeronasal 1 receptor 86
- Gene Aliases
- ENSMUSG00000070816, Gm10301
- Chromosome Location
- Chr. 7: 13101997 - 13102947 on Build GRCm38
- UniGene ID
- Mm.480296
- Species
- Mus musculus
- Gene Symbol
- Vmn1r88
- Entrez Gene ID
- 100312474
- Gene Name
- vomeronasal 1 receptor, 88
- Gene Aliases
- Chromosome Location
- Chr. 7: 13177719 - 13178669 on Build GRCm38
- UniGene ID
- Mm.480295
- Species
- Mus musculus
View Full Product Details