Gene Transcripts

Gene Symbol
Man2a1
Entrez Gene ID
17158
Gene Name
mannosidase 2, alpha 1
Gene Aliases
Mana-2, Mana2, Map-2
Chromosome Location
Chr. 17: 64601649 - 64755110 on Build GRCm38
UniGene ID
Mm.2433
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008549.2 NP_032575.2 14 2392
GenBank mRNA BC138371.1 - 14 2377
BC138372.1 - 14 2377 8861392
X61172.1 - 14 2392

View Full Product Details