Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008613.3 | NP_032639.1 | 4, 5 | 611 | |
GenBank mRNA | AK013048.1 | - | 4, 5 | 578 | |
AK132772.1 | - | 4, 5 | 586 | ||
BC046624.1 | - | 4, 5 | 524 | 6314904 | |
D14849.1 | - | 4, 5 | 642 |