Gene Transcripts

Gene Symbol
Mdh2
Entrez Gene ID
17448
Gene Name
malate dehydrogenase 2, NAD (mitochondrial)
Gene Aliases
MDH, Mdh-2, Mor-1, Mor1
Chromosome Location
Chr. 5: 135778649 - 135790386 on Build GRCm38
UniGene ID
Mm.297096
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008617.2 NP_032643.2 8 936
GenBank mRNA AK002305.1 - 8 953
AK135162.1 - 8 949
AK160553.1 - 8 919
AK167809.1 - 8 919
BC023482.1 - 8 936 5251192
DQ402950.1 - 7 786
M16229.1 - 8 895

View Full Product Details