Gene Transcripts
- Gene Symbol
- Myb
- Entrez Gene ID
- 17863
- Gene Name
- myeloblastosis oncogene
- Gene Aliases
- AI550390, M16449, c-myb
- Chromosome Location
- Chr. 10: 21124930 - 21160984 on Build GRCm38
- UniGene ID
- Mm.473872
- Species
- Mus musculus
- Gene Symbol
- LOC101056071
- Entrez Gene ID
- 101056071
- Gene Name
- uncharacterized LOC101056071
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.450591
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138601.1 |
- |
2 |
342 |
|
View Full Product Details