Gene Transcripts

Gene Symbol
Mybpc3
Entrez Gene ID
17868
Gene Name
myosin binding protein C, cardiac
Gene Aliases
Chromosome Location
Chr. 2: 91118114 - 91136518 on Build GRCm38
UniGene ID
Mm.10728
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008653.2 NP_032679.2 33 3730
GenBank mRNA AF097333.1 - 33 3681
AK146661.1 - 33 3731
AK146886.1 - 33 3731
AK169302.1 - 32 3708
AK169306.1 - 32 3708
BC094945.1 - 33 3727
BC131672.1 - 33 3704
BC154408.1 - 33 3751 40110890

View Full Product Details