Gene Transcripts

Gene Symbol
Odf2
Entrez Gene ID
18286
Gene Name
outer dense fiber of sperm tails 2
Gene Aliases
AI848335, MMTEST29
Chromosome Location
Chr. 2: 29889720 - 29931746 on Build GRCm38
UniGene ID
Mm.330116
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113213.1 NP_001106684.1 5 653
NM_001113214.1 NP_001106685.1 5 653
NM_001177659.1 NP_001171130.1 5 653
NM_001177661.1 NP_001171132.1 4 499
NM_013615.3 NP_038643.1 5 626
GenBank mRNA AF000968.1 - 3 473
AF034105.1 - 5 589
AK143726.1 - 5 557
AK148523.1 - 5 566
AK165588.1 - 5 557
AK168415.1 - 5 555
BC057001.1 - 6 710 6827260
BQ840288.1 - 3 211

View Full Product Details