Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011053.2 | NP_035183.2 | 24 | 3803 | |
GenBank mRNA | AK129080.1 | - | 24 | 3795 | |
AK141450.1 | - | 24 | 3804 | ||
AK147950.1 | - | 24 | 3788 | ||
BC010844.1 | - | 24 | 3778 | ||
BC070468.1 | - | 24 | 3790 | 30628812 | |
BC094244.1 | - | 5 | 844 |