Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011084.2 | NP_035214.2 | 12 | 1732 | |
NM_207683.2 | NP_997566.1 | 4478 | |||
GenBank mRNA | AB008791.1 | - | 4478 | ||
AB008792.1 | - | 12 | 1727 | ||
BC150813.1 | - | 4470 | 9087724 |