Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133796.7 | NP_598557.3 | 4 | 443 | |
GenBank mRNA | AK075656.1 | - | 3 | 310 | |
AK077343.1 | - | 4 | 603 | ||
AK143516.1 | - | 4 | 427 | ||
AK147842.1 | - | 402 | |||
AK156819.1 | - | 388 | |||
AK165241.1 | - | 4 | 425 | ||
AK170553.1 | - | 4 | 410 | ||
AK170718.1 | - | 4 | 427 | ||
AK172140.1 | - | 388 | |||
BC004732.1 | - | 4 | 385 | 3583026 | |
BC086755.1 | - | 4 | 367 | 6414768 |