Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011340.3 | NP_035470.3 | 6 | 887 | |
GenBank mRNA | AF017057.1 | - | 6 | 785 | |
AF036164.1 | - | 6 | 785 | ||
AK219600.1 | - | 11 | |||
BC019852.1 | - | 6 | 797 | 5123884 | |
D50460.1 | - | 6 | 856 | ||
D87975.1 | - | 6 | 784 |