Gene Transcripts

Gene Symbol
Smarcc1
Entrez Gene ID
20588
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1
Gene Aliases
AI115498, BAF155, Rsc8, SRG3
Chromosome Location
Chr. 9: 110132024 - 110240178 on Build GRCm38
UniGene ID
Mm.85410
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009211.2 NP_033237.2 7, 8 816
GenBank mRNA AK042194.1 - 8, 9 729
AK133922.1 - 7, 8 791
AK143485.1 - 7, 8 804
AK144122.1 - 7, 8 790
AK144631.1 - 7, 8 784
AK145352.1 - 7, 8 783
BC022173.1 - 7, 8 780
BC052423.1 - 7, 8 778 6400646
BC053064.1 - 7, 8 772
BC056375.1 - 7, 8 779
U85614.1 - 7, 8 777

View Full Product Details