Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011461.3 | NP_035591.3 | 6 | 520 | |
GenBank mRNA | AF098863.2 | - | 6 | 519 | |
AK137522.1 | - | 6 | 523 | ||
AK156024.1 | - | 6 | 521 | ||
AK156892.1 | - | 6 | 521 | ||
AK157188.1 | - | 7 | 615 | ||
BC064038.1 | - | 5 | 481 | 3824951 |