Gene Transcripts

Gene Symbol
Srm
Entrez Gene ID
20810
Gene Name
spermidine synthase
Gene Aliases
AA407669, SpdST, SpdSy
Chromosome Location
Chr. 4: 148591513 - 148594619 on Build GRCm38
UniGene ID
Mm.10
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009272.4 NP_033298.1 4 613
GenBank mRNA AK028359.1 - 4 624
AK051101.1 - 4 609
AK146395.1 - 4 570
BC005566.1 - 4 617 3966710
L19311.1 - 4 581

View Full Product Details