Gene Transcripts

Gene Symbol
Enox2
Entrez Gene ID
209224
Gene Name
ecto-NOX disulfide-thiol exchanger 2
Gene Aliases
APK1, Cova1, tNOX
Chromosome Location
Chr. X: 49009707 - 49288245 on Build GRCm38
UniGene ID
Mm.327730
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271447.1 NP_001258376.1 14 1935
NM_001271448.1 NP_001258377.1 13 1780
NM_001271449.1 NP_001258378.1 14 1924
NM_001271450.1 NP_001258379.1 14 1891
NM_001271451.1 NP_001258380.1 14 1924
NM_145951.5 NP_666063.1 14 1879
GenBank mRNA AK028343.1 - 14 1929
AK034058.1 - 14 1880
AK041822.1 - 14 1892
AK133956.1 - 13 1751
AK137736.1 - 14 1906
AK139897.1 - 14 1906
BC025915.1 - 14 1898 5043650
BC026450.1 - 6 571

View Full Product Details