Gene Transcripts

Gene Symbol
Chtf8
Entrez Gene ID
214987
Gene Name
CTF8, chromosome transmission fidelity factor 8
Gene Aliases
5830457O10Rik, Ctf8, Derpc
Chromosome Location
Chr. 8: 106883863 - 106893593 on Build GRCm38
UniGene ID
Mm.490308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145412.3 NP_663387.3 4 533
GenBank mRNA AK030924.1 - 4 532
AK088257.1 - 4 542
AK165622.1 - 4 515
AK165993.1 - 2 359
BC023107.1 - 2 305 3966700

View Full Product Details