Gene Transcripts

Gene Symbol
Bud13
Entrez Gene ID
215051
Gene Name
BUD13 homolog (yeast)
Gene Aliases
AV305342, D030060M11Rik
Chromosome Location
Chr. 9: 46283012 - 46298783 on Build GRCm38
UniGene ID
Mm.32648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146000.2 NP_666112.1 1, 2 170
GenBank mRNA AK037134.1 - 1, 2 167
AK038126.1 - 1, 2 175
AK051047.1 - 1, 2 175
AK077744.1 - 1, 2 175
BC025490.1 - 1, 2 171 5252666

View Full Product Details