Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164376.1 | NP_001157848.1 | 18 | 2723 | |
NM_001164519.1 | NP_001157991.1 | 13 | 1882 | ||
NM_177612.3 | NP_808280.2 | 18 | 2651 | ||
GenBank mRNA | AF344871.1 | - | 18 | 2723 | |
AK030166.1 | - | 18 | 2652 | ||
AK142745.1 | - | 18 | 2724 | ||
BC112409.1 | - | 18 | 2625 | 40060582 | |
DQ380429.1 | - | 18 | 2651 | ||
DQ380430.1 | - | 13 | 1881 |