Gene Transcripts
- Gene Symbol
- Ilvbl
- Entrez Gene ID
- 216136
- Gene Name
- ilvB (bacterial acetolactate synthase)-like
- Gene Aliases
- 5830463I21, AI415009
- Chromosome Location
- Chr. 10: 78574345 - 78584502 on Build GRCm38
- UniGene ID
- Mm.2644
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173751.4 |
NP_776112.1
|
6, 7 |
885 |
|
GenBank mRNA |
AK030933.1 |
- |
3, 4 |
323 |
|
AK087956.1 |
- |
5, 6 |
759 |
|
AK133553.1 |
- |
5, 6 |
815 |
|
AK138031.1 |
- |
5, 6 |
734 |
|
AK143918.1 |
- |
5, 6 |
784 |
|
AK146075.1 |
- |
5, 6 |
813 |
|
AK169899.1 |
- |
5, 6 |
811 |
|
BC052054.1 |
- |
5, 6 |
1219 |
|
BC056459.1 |
- |
6, 7 |
843 |
5718157 |
View Full Product Details