Gene Transcripts

Gene Symbol
Otop1
Entrez Gene ID
21906
Gene Name
otopetrin 1
Gene Aliases
A530025J20Rik, Otp1, tlt
Chromosome Location
Chr. 5: 38277404 - 38304217 on Build GRCm38
UniGene ID
Mm.204765
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172709.3 NP_766297.2 6 1717
GenBank mRNA AF548337.1 - 6 1717
AK040792.1 - 6 1704
AK041023.1 - 6 1491

View Full Product Details