Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021885.4 | NP_068685.1 | 7 | 1076 | |
GenBank mRNA | AK147700.1 | - | 7 | 1131 | |
AK165720.1 | - | 7 | 1077 | ||
BC096554.1 | - | 7 | 1057 | 30616697 | |
U52433.1 | - | 7 | 1064 | ||
U52824.1 | - | 8 | 1209 | ||
U54643.1 | - | 7 | 994 |