Gene Transcripts

Gene Symbol
E430025E21Rik
Entrez Gene ID
223593
Gene Name
RIKEN cDNA E430025E21 gene
Gene Aliases
AL022848, C76463, Kiaa0196, mKIAA0196, strumpellin
Chromosome Location
Chr. 15: 59331997 - 59374194 on Build GRCm38
UniGene ID
Mm.218665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153548.2 NP_705776.2 25 3249
GenBank mRNA AK088754.1 - 10 1122
AK128970.1 - 25 3264
AK129086.1 - 2901
AK163968.1 - 8 873
BC031364.1 - 11 1263
BC034070.1 - 15 1849 4217923
BC040815.1 - 15 1849 4217923
BC067035.1 - 25 3249 30532658

View Full Product Details