Gene Transcripts

Gene Symbol
Arhgap30
Entrez Gene ID
226652
Gene Name
Rho GTPase activating protein 30
Gene Aliases
6030405P05Rik, Gm102, mFLJ00267
Chromosome Location
Chr. 1: 171388960 - 171410240 on Build GRCm38
UniGene ID
Mm.251048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005508.2 NP_001005508.2 12 2021
GenBank mRNA AK031319.1 - 12 1792
AK147757.1 - 11 2121
AK156351.1 - 12 2035
AK220204.1 - 7 1179
BC082573.1 - 12 1969 30548035

View Full Product Details