Gene Transcripts

Gene Symbol
Ccnyl1
Entrez Gene ID
227210
Gene Name
cyclin Y-like 1
Gene Aliases
9630037P07Rik, AW554339
Chromosome Location
Chr. 1: 64691005 - 64725646 on Build GRCm38
UniGene ID
Mm.39706
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097644.1 NP_001091113.1 10 869
GenBank mRNA AK132782.1 - 1224
DV657169.1 - 11

View Full Product Details