Gene Transcripts
- Gene Symbol
- Dgkd
- Entrez Gene ID
- 227333
- Gene Name
- diacylglycerol kinase, delta
- Gene Aliases
- AI841987, D330025K09, DGKdelta, dgkd-2
- Chromosome Location
- Chr. 1: 87853273 - 87944490 on Build GRCm38
- UniGene ID
- Mm.277217
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177646.3 |
NP_808314.2
|
19 |
2334 |
|
GenBank mRNA |
AK172901.1 |
- |
15 |
1779 |
|
View Full Product Details