Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012392.1 | NP_001012392.1 | 11, 12 | 1256 | |
NM_153418.2 | NP_700467.2 | 11, 12 | 1219 | ||
GenBank mRNA | AK146605.1 | - | 11, 12 | 1252 | |
BC060982.1 | - | 11, 12 | 1256 | 30300112 | |
BC100436.1 | - | 11, 12 | 1251 | 30298932 | |
U46068.3 | - | 11, 12 | 1220 |