Gene Transcripts
- Gene Symbol
- Spg20
- Entrez Gene ID
- 229285
- Gene Name
- spastic paraplegia 20, spartin (Troyer syndrome) homolog (human)
- Gene Aliases
- AI840044, C79168, SPARTIN, TAHCCP1, mKIAA0610
- Chromosome Location
- Chr. 3: 55112074 - 55137332 on Build GRCm38
- UniGene ID
- Mm.235523
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001144987.1 |
NP_001138459.1
|
4, 5 |
1517 |
|
NM_001144988.1 |
NP_001138460.1
|
3, 4 |
1130 |
|
NM_144895.2 |
NP_659144.1
|
4, 5 |
1301 |
|
GenBank mRNA |
AK031545.1 |
- |
3, 4 |
856 |
|
AK035081.1 |
- |
2, 3 |
989 |
|
AK036569.1 |
- |
2, 3 |
453 |
|
AK087388.1 |
- |
4, 5 |
1270 |
|
AK089946.1 |
- |
4, 5 |
1302 |
|
AK129171.1 |
- |
3, 4 |
1109 |
|
AK147760.1 |
- |
4, 5 |
1519 |
|
AK159951.1 |
- |
4, 5 |
1302 |
|
AK192376.1 |
- |
2, 3 |
203 |
|
AK196543.1 |
- |
1, 2 |
37 |
|
AK200807.1 |
- |
1, 2 |
37 |
|
BC022921.1 |
- |
4, 5 |
1269 |
4220506 |
View Full Product Details