Gene Transcripts

Gene Symbol
Gpr61
Entrez Gene ID
229714
Gene Name
G protein-coupled receptor 61
Gene Aliases
Chromosome Location
Chr. 3: 108148321 - 108154986 on Build GRCm38
UniGene ID
Mm.73666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK032627.1 - 2 850
AK133702.1 - 2 849
BC116333.2 - 1 506 40096913
BC116334.2 - 1 506 40096914

View Full Product Details