Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172687.1 | NP_766275.1 | 7 | 1069 | |
GenBank mRNA | AK028680.1 | - | 7 | 1069 | |
AK035318.1 | - | 7 | 1064 | ||
AK142287.1 | - | 7 | 1007 | ||
AK166475.1 | - | 7 | 1024 | ||
AK170488.1 | - | 7 | 1064 | ||
BC060960.1 | - | 7 | 1071 | 30312962 |