Gene Transcripts

Gene Symbol
Slc30a2
Entrez Gene ID
230810
Gene Name
solute carrier family 30 (zinc transporter), member 2
Gene Aliases
6030430C21, ZnT-2, Znt2
Chromosome Location
Chr. 4: 134342961 - 134356528 on Build GRCm38
UniGene ID
Mm.358876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039677.2 NP_001034766.1 6, 7 1067
GenBank mRNA AK031425.1 - 6, 7 863
BC013478.1 - 5, 6 1087
BC113751.1 - 6, 7 1067 40057211
BC147704.1 - 6, 7 898 9087463
BC147707.1 - 6, 7 898 9087466

View Full Product Details