Gene Transcripts

Gene Symbol
Cc2d2a
Entrez Gene ID
231214
Gene Name
coiled-coil and C2 domain containing 2A
Gene Aliases
5730509K17Rik, b2b1035Clo
Chromosome Location
Chr. 5: 43662373 - 43740972 on Build GRCm38
UniGene ID
Mm.44434
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172274.2 NP_758478.1 38 4952
GenBank mRNA AK037073.1 - 25 3354
BC032290.1 - 38 4896 5369681

View Full Product Details