Gene Transcripts

Gene Symbol
Ddx60
Entrez Gene ID
234311
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 60
Gene Aliases
9830118M07
Chromosome Location
Chr. 8: 61928066 - 62037704 on Build GRCm38
UniGene ID
Mm.275233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293783.1 NP_001280712.1 36 5208
GenBank mRNA AK136403.1 - 3 2288
BC013672.1 - 9 1196

View Full Product Details