Gene Transcripts
- Gene Symbol
- Inpp4b
- Entrez Gene ID
- 234515
- Gene Name
- inositol polyphosphate-4-phosphatase, type II
- Gene Aliases
- E130107I17Rik
- Chromosome Location
- Chr. 8: 81342342 - 82152019 on Build GRCm38
- UniGene ID
- Mm.324468
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001024617.3 |
NP_001019788.1
|
14, 15 |
1612 |
|
NM_001297591.1 |
NP_001284520.1
|
14, 15 |
1589 |
|
NM_001297593.1 |
NP_001284522.1
|
13, 14 |
1470 |
|
NM_001297596.1 |
NP_001284525.1
|
14, 15 |
1589 |
|
GenBank mRNA |
AK053554.1 |
- |
7, 8 |
741 |
|
AK089068.1 |
- |
9, 10 |
843 |
|
AK139397.1 |
- |
7, 8 |
1089 |
|
AK141502.1 |
- |
16, 17 |
1586 |
|
BC064813.1 |
- |
12, 13 |
1314 |
6826283 |
DQ296611.1 |
- |
14, 15 |
1589 |
|
DQ296612.1 |
- |
13, 14 |
1470 |
|
DQ296613.1 |
- |
14, 15 |
1589 |
|
View Full Product Details