Gene Transcripts

Gene Symbol
Ttc13
Entrez Gene ID
234875
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
BC017545
Chromosome Location
Chr. 8: 124671327 - 124722013 on Build GRCm38
UniGene ID
Mm.38436
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145607.3 NP_663582.2 15 1732
GenBank mRNA AK036992.1 - 1 3651
AK144842.1 - 15 1733
AK148102.1 - 17 1962
AK160894.1 - 17 1969
BC017545.1 - 9 943
BC072577.1 - 15 1722
BC079851.1 - 16 1787 6401930

View Full Product Details