Gene Transcripts
- Gene Symbol
- Trim43b
- Entrez Gene ID
- 666747
- Gene Name
- tripartite motif-containing 43B
- Gene Aliases
- EG666747, Gm8269
- Chromosome Location
- Chr. 9: 89084624 - 89092835 on Build GRCm38
- UniGene ID
- Mm.425592
- Species
- Mus musculus
- Gene Symbol
- Trim43c
- Entrez Gene ID
- 666731
- Gene Name
- tripartite motif-containing 43C
- Gene Aliases
- EG666731, Trim43
- Chromosome Location
- Chr. 9: 88839023 - 88848198 on Build GRCm38
- UniGene ID
- Mm.446951
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DV649510.1 |
- |
|
448 |
|
Chr. - - [-] |
View Full Product Details