Gene Transcripts

Gene Symbol
Aoc2
Entrez Gene ID
237940
Gene Name
amine oxidase, copper containing 2 (retina-specific)
Gene Aliases
RAO
Chromosome Location
Chr. 11: 101325063 - 101329699 on Build GRCm38
UniGene ID
Mm.302277
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178932.1 NP_849263.1 3 1952
GenBank mRNA AF350445.2 - 3 1952
BC150843.1 - 3 1943 9087754
BC150848.1 - 3 1943 9087759

View Full Product Details