Gene Transcripts

Gene Symbol
Cdh24
Entrez Gene ID
239096
Gene Name
cadherin-like 24
Gene Aliases
1700040A22Rik, ENSMUSG00000022188
Chromosome Location
Chr. 14: 54631992 - 54641364 on Build GRCm38
UniGene ID
Mm.386871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199470.2 NP_955764.1 10 1838
GenBank mRNA BC057373.1 - 1792

View Full Product Details