Gene Transcripts

Gene Symbol
Cacna1i
Entrez Gene ID
239556
Gene Name
calcium channel, voltage-dependent, alpha 1I subunit
Gene Aliases
Ca(V)3.3
Chromosome Location
Chr. 15: 80287238 - 80398292 on Build GRCm38
UniGene ID
Mm.291058
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044308.2 NP_001037773.2 14, 15 3047
GenBank mRNA AK147585.1 - 14, 15 3047

View Full Product Details