Gene Transcripts

Gene Symbol
Carf
Entrez Gene ID
241066
Gene Name
calcium response factor
Gene Aliases
Als2cr8, ECBRC-FC1, ECBRC-FC2
Chromosome Location
Chr. 1: 60098221 - 60153953 on Build GRCm38
UniGene ID
Mm.276441
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285463.1 NP_001272392.1 17 2269
NM_001285473.1 NP_001272402.1 13 1859
NM_139150.5 NP_631889.1 15 2142
GenBank mRNA AB221600.1 - 14 1925
AF454947.1 - 15 2111
AK044738.1 - 17 2270
BC138644.1 - 14 2048 8861664
BC138645.1 - 14 2048 8861665
BC145350.1 - 14 2038
EU170502.1 - 13 1840
EU170503.1 - 11 1575

View Full Product Details