Gene Transcripts

Gene Symbol
Doxl2
Entrez Gene ID
243376
Gene Name
diamine oxidase-like protein 2
Gene Aliases
Chromosome Location
Chr. 6: 48973816 - 48978745 on Build GRCm38
UniGene ID
Mm.244112
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029987.1 NP_001025158.1 1 45
GenBank mRNA BC150935.1 - 1 114

View Full Product Details