Gene Transcripts

Gene Symbol
Nanos3
Entrez Gene ID
244551
Gene Name
nanos homolog 3 (Drosophila)
Gene Aliases
nos3
Chromosome Location
Chr. 8: 84173730 - 84176945 on Build GRCm38
UniGene ID
Mm.216740
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194059.2 NP_918948.1 1, 2 483
GenBank mRNA AB095973.1 - 1, 2 483
BC119513.1 - 1, 2 483 40055381

View Full Product Details