Gene Transcripts

Gene Symbol
Pskh1
Entrez Gene ID
244631
Gene Name
protein serine kinase H1
Gene Aliases
AW539964, E130013P03Rik, b2b1230Clo
Chromosome Location
Chr. 8: 105900474 - 105931802 on Build GRCm38
UniGene ID
Mm.27627
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173432.2 NP_775608.1 2, 3 1101
GenBank mRNA AK053397.1 - 2, 3 1135
AK153861.1 - 2, 3 1148
AK154570.1 - 2, 3 1131
AK166338.1 - 2, 3 1125
AK191303.1 - 1, 2 195
BC050128.1 - 2, 3 1101 30024345

View Full Product Details