Gene Transcripts

Gene Symbol
Abcg2
Entrez Gene ID
26357
Gene Name
ATP-binding cassette, sub-family G (WHITE), member 2
Gene Aliases
ABC15, ABCP, AI428558, BCRP, Bcrp1, MXR, MXR1
Chromosome Location
Chr. 6: 58584491 - 58692451 on Build GRCm38
UniGene ID
Mm.333096
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011920.3 NP_036050.1 9, 10 1646
GenBank mRNA AF140218.1 - 8, 9 1209
BC053730.1 - 9, 10 1658 30064535
KF662864.1 - 9, 10 1243

View Full Product Details