Gene Transcripts

Gene Symbol
Plod3
Entrez Gene ID
26433
Gene Name
procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3
Gene Aliases
AI414586, LH3
Chromosome Location
Chr. 5: 136987019 - 136996646 on Build GRCm38
UniGene ID
Mm.251003
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011962.3 NP_036092.1 13 2046
GenBank mRNA AF046783.1 - 13 1454
AK013195.1 - 13 1592
AK033360.1 - 13 1591
AK053320.1 - 13 1688
AK088948.1 - 13 1463
BC043047.1 - 13 2045 5703967
BC054734.1 - 13 2046 5720686

View Full Product Details