Gene Transcripts
- Gene Symbol
- Abca13
- Entrez Gene ID
- 268379
- Gene Name
- ATP-binding cassette, sub-family A (ABC1), member 13
- Gene Aliases
- 9830132L24, A930002G16Rik, AI956815
- Chromosome Location
- Chr. 11: 9191354 - 9684871 on Build GRCm38
- UniGene ID
- Mm.39270
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178259.3 |
NP_839990.2
|
50 |
13390 |
|
GenBank mRNA |
AY160971.1 |
- |
32 |
4709 |
|
View Full Product Details