Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178749.3 | NP_848864.1 | 11, 12 | 1271 | |
GenBank mRNA | AK036266.1 | - | 11, 12 | 1272 | |
AK042599.1 | - | 11, 12 | 1385 | ||
AK044474.1 | - | 11, 12 | 1281 | ||
BC055002.1 | - | 1201 | 6313304 |