Gene Transcripts

Gene Symbol
Ttll13
Entrez Gene ID
269954
Gene Name
tubulin tyrosine ligase-like family, member 13
Gene Aliases
1700111A04Rik, 4921538O11, BC024578
Chromosome Location
Chr. 7: 80246376 - 80260821 on Build GRCm38
UniGene ID
Mm.276780
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177765.3 NP_808433.2 14 2500
GenBank mRNA AK029565.1 - 14 2499
AM690757.1 - 13 2201
BC145749.1 - 13 2108

View Full Product Details