Gene Transcripts

Gene Symbol
Klhdc1
Entrez Gene ID
271005
Gene Name
kelch domain containing 1
Gene Aliases
Chromosome Location
Chr. 12: 69226917 - 69284646 on Build GRCm38
UniGene ID
Mm.180969
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178253.5 NP_839984.1 10 956
GenBank mRNA AK144621.1 - 10 896
AY238938.1 - 10 956
BC119567.2 - 10 934 40057793
BC119568.2 - 10 940 40057794

View Full Product Details