Gene Transcripts

Gene Symbol
Prss43
Entrez Gene ID
272643
Gene Name
protease, serine 43
Gene Aliases
Gm661, Tessp3
Chromosome Location
Chr. 9: 110826690 - 110831507 on Build GRCm38
UniGene ID
Mm.458525
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199471.1 NP_955765.1 4 1008
GenBank mRNA AB100999.1 - 4 1008

View Full Product Details